Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2,6-Trimethyl-6-vinyltetrahydropyran-3-ol (mixture of isomers) 98.0+%, TCI America™
SDP

Catalog No. T260525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T260525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T260525G Supplier TCI America Supplier No. T260525G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14049-11-7
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD06200770
InChI Key BCTBAGTXFYWYMW-UHFFFAOYSA-N
Synonym 3-Hydroxy-2,2,6-trimethyl-6-vinyltetrahydropyran, Linalool Oxide Pyranoid
PubChem CID 26396
IUPAC Name 6-ethenyl-2,2,6-trimethyloxan-3-ol
SMILES CC1(C(CCC(O1)(C)C=C)O)C

Specifications

Color Colorless
Boiling Point 98°C
Quantity 25 g
Formula Weight 170.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,2,6-Trimethyl-6-vinyltetrahydropyran-3-ol (mixture of isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.