accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14059-33-7

IUPAC Name: bismuth(3+) vanadium tetraoxidandiide
Synonyms: bismuth vanadate
Molecular Weight (g/mol): 323.92
Molecular Formula: BiO4V
InChi Key: HUUOUJVWIOKBMD-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[O--].[V].[Bi+3]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results