Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bismuth vanadium oxide, 99.9% (metals basis)

Catalog No. AA3922022
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3922022 100 g
AA3922014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3922022 Supplier Thermo Scientific Chemicals Supplier No. 03922022
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 14059-33-7 | BiO4V | 323.918 g/mol

Bismuth vanadium oxide is used as a pigment due to its hiding power. It also finds use in paints, coatings, plastic and rubber products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14059-33-7
Molecular Formula BiO4V
Molecular Weight (g/mol) 323.918
MDL Number MFCD00064836
InChI Key YNOWMDYNXXVJMC-UHFFFAOYSA-N
Synonym Bismuth vanadate
PubChem CID 20243764
IUPAC Name bismuth;trioxido(oxo)vanadium
SMILES [O-][V](=O)([O-])[O-].[Bi+3]

Specifications

Color Red to Yellow
Physical Form Powder
Quantity 100 g
UN Number UN3285
Solubility Information Soluble in acids; Insoluble in water.
Formula Weight 323
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Bismuth vanadium oxide
Hazard Category Warning
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: VANADIUM COMPOUND, N.O.S.
EINECSNumber 237-898-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.