accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNNNCH3CH2OOCH3CH3CH3CH3CH3CH3OOCH3OH3CCH3Mg(E/Z)(E/Z)(E/Z)(E/Z)

CAS RN 1406-65-1

IUPAC Name: magnesium(2+) 16-ethenyl-11-ethyl-3-(methoxycarbonyl)-12,17,21,26-tetramethyl-22-{3-oxo-3-[(3,7,11,15-tetramethylhexadec-2-en-1-yl)oxy]propyl}-7,23,24,25-tetraazahexacyclo[18.2.1.1⁵,⁸.1¹⁰,¹³.1¹⁵,¹⁸.0²,⁶]hexacosa-1(23),2(6),4,8(26),9,11,13(25),14,16,18(24),19-undecaen-7-id-4-olate
Synonyms: magnesium(2+) 16-ethenyl-11-ethyl-3-(methoxycarbonyl)-12,17,21,26-tetramethyl-22-{3-oxo-3-[(3,7,11,15-tetramethylhexadec-2-en-1-yl)oxy]propyl}-7,23,24,25-tetraazahexacyclo[18.2.1.1⁵,⁸.1¹⁰,¹³.1¹⁵,¹⁸.0²,⁶]hexacosa-1(23),2(6),4,8(26),9,11,13(25),14,16,18(24),19-undecaen-7-id-4-olate
Molecular Weight (g/mol): 893.51
Molecular Formula: C55H72MgN4O5
InChi Key: ATNHDLDRLWWWCB-UHFFFAOYNA-M
SMILES: [Mg++].CCC1=C(C)C2=NC1=CC1=C(C)C3=C([O-])C(C(=O)OC)C(=C3[N-]1)C1=NC(=CC3=NC(=C2)C(C=C)=C3C)C(C)C1CCC(=O)OCC=C(C)CCCC(C)CCCC(C)CCCC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results