Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorophyll, Water Soluble, MP Biomedicals

Catalog No. ICN210221 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN210221 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN210221 Supplier MP Biomedicals, Inc Supplier No. 0521022180
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1406-65-1
Molecular Formula C55H72MgN4O5
Molecular Weight (g/mol) 893.509
InChI Key ATNHDLDRLWWWCB-AENOIHSZSA-M
Synonym unii-yf5q9ejc8y, chlorophyll a, chlorophyll-a, yf5q9ejc8y, chlorophyll, magnesium, 2e,7r,11r-3,7,11,15-tetramethyl-2-hexadecenyl 3s,4s,21r-9-ethenyl-14-ethyl-21-methoxycarbonyl-4,8,13,18-tetramethyl-20-oxo-3-phorbinepropanoato 2--kappan23,kappan24,kappan25,kappan26-, sp-4-2, unii-00wnz48or9 component, 2s-1-4r,7s,10s,13s,16s,19r-19-amino-7-2-amino-2-oxo-ethyl-10-3-amino-3-oxo-propyl-13-benzyl-16-4-hydroxyphenyl methyl-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentazacycloicosane-4-carbonyl-n-1s-1-2-amino-2-oxo-ethyl carbamoyl-4-guanidino-butyl pyrro, 2s-1-4r,7s,10s,13s,16s,19r-19-amino-7-2-amino-2-oxo-ethyl-10-3-amino-3-oxo-propyl-13-benzyl-16-4-hydroxyphenyl methyl-6,9,12,15,18-pentaoxo-1,2-dithia-5,8,11,14,17-pentazacycloicosane-4-carbonyl-n-1s-5-amino-1-2-amino-2-oxo-ethyl carbamoyl pentyl pyrrolid, magnesium, 3,7,11,15-tetramethyl-2-hexadecenyl 9-ethenyl-14-ethyl-21-methoxycarbonyl-4,8,13,18-tetramethyl-20-oxo-3-phorbinepropanoato 2--n23,n24,n25,n26-, sp-4-2-3s-3alpha 2e,7s*,11s* ,4beta,21beta
PubChem CID 16667503
ChEBI CHEBI:18230
SMILES CCC1=C(C2=NC1=CC3=C(C4=C(C(C(=C4[N-]3)C5=NC(=CC6=NC(=C2)C(=C6C)C=C)C(C5CCC(=O)OCC=C(C)CCCC(C)CCCC(C)CCCC(C)C)C)C(=O)OC)[O-])C)C.[Mg+2]

Specifications

Quantity 100 g
Chemical Name or Material Chlorophyll, Water Soluble
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.