accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHNNNHNNS

CAS RN 14070-48-5

IUPAC Name: N-[3-(5-sulfanylidene-2,5-dihydro-1H-1,2,3,4-tetrazol-1-yl)phenyl]acetamide
Synonyms: N-[3-(5-sulfanylidene-2H-1,2,3,4-tetrazol-1-yl)phenyl]acetamide
Molecular Weight (g/mol): 235.27
Molecular Formula: C9H9N5OS
InChi Key: SCWKACOBHZIKDI-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=CC(=C1)N1NN=NC1=S
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results