Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Acetamidophenyl)-5-mercaptotetrazole 98.0+%, TCI America™
SDP

Catalog No. A135210G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A135210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A135210G Supplier TCI America Supplier No. A135210G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14070-48-5
Molecular Formula C9H9N5OS
Molecular Weight (g/mol) 235.27
MDL Number MFCD00603728
InChI Key SCWKACOBHZIKDI-UHFFFAOYSA-N
PubChem CID 712430
IUPAC Name N-[3-(5-sulfanylidene-2,5-dihydro-1H-1,2,3,4-tetrazol-1-yl)phenyl]acetamide
SMILES CC(=O)NC1=CC=CC(=C1)N1NN=NC1=S

Specifications

Color White-Yellow
Quantity 10 g
UN Number 1325
Formula Weight 235.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(3-Acetamidophenyl)-5-mercaptotetrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.