accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONONH2

CAS RN 140836-69-7

IUPAC Name: 1-{4-[2-(morpholin-4-yl)ethoxy]phenyl}methanamine
Synonyms: 1-{4-[2-(morpholin-4-yl)ethoxy]phenyl}methanamine
Molecular Weight (g/mol): 236.32
Molecular Formula: C13H20N2O2
InChi Key: KMSJKJIIAVTIKY-UHFFFAOYSA-N
SMILES: NCC1=CC=C(OCCN2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results