accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 141-18-4

IUPAC Name: 1,6-bis(2-butoxyethyl) hexanedioate
Synonyms: 1,6-bis(2-butoxyethyl) hexanedioate
Molecular Weight (g/mol): 346.46
Molecular Formula: C18H34O6
InChi Key: IHTSDBYPAZEUOP-UHFFFAOYSA-N
SMILES: CCCCOCCOC(=O)CCCCC(=O)OCCOCCCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results