accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 141-57-1

IUPAC Name: trichloro(propyl)silane
Synonyms: propyltrichlorosilane
Molecular Weight (g/mol): 177.52
Molecular Formula: C3H7Cl3Si
InChi Key: DOEHJNBEOVLHGL-UHFFFAOYSA-N
SMILES: CCC[Si](Cl)(Cl)Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results