Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloro(propyl)silane 99.0+%, TCI America™
SDP

Catalog No. P037125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P037125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P037125G Supplier TCI America Supplier No. P037125G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 141-57-1
Molecular Formula C3H7Cl3Si
Molecular Weight (g/mol) 177.524
MDL Number MFCD00013603
InChI Key DOEHJNBEOVLHGL-UHFFFAOYSA-N
Synonym propyltrichlorosilane, trichloro propyl silane, n-propyltrichlorosilane, silane, trichloropropyl, propyl trichlorosilane, trichloro-n-propylsilane, silane, propyltrichloro, unii-x16g85fi4y, hsdb 889, silane,trichloropropyl
PubChem CID 8850
IUPAC Name trichloro(propyl)silane
SMILES CCC[Si](Cl)(Cl)Cl

Specifications

Color Yellow
Boiling Point 124°C
Quantity 25 g
UN Number 1816
Formula Weight 177.52
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Trichloro(propyl)silane
EINECSNumber (2)-2041
RTECSNumber VV5300000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.