accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 141022-99-3

IUPAC Name: 1,1,1-trifluoroheptadecan-2-one
Synonyms: 1,1,1-trifluoroheptadecan-2-one
Molecular Weight (g/mol): 308.43
Molecular Formula: C17H31F3O
InChi Key: MAHYXYTYTLCTQD-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCC(=O)C(F)(F)F
Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results