Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ PACOCF3

Catalog No. 5062745MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-627-45MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-627-45MG Supplier MilliporeSigma™ Supplier No. 5062745MG
Add to Cart
Add to Cart

A novel Ca2+-independent and reversible inhibitor of phospholipase A2 (PLA2; IC50 = 3.8 μM).

A novel Ca2+-independent and reversible inhibitor of phospholipase A2 (PLA2; IC50 = 3.8μM). May also inhibit Ca2+-dependent PLA2 at much higher concentrations (IC50 = 45μM).
TRUSTED_SUSTAINABILITY

Specifications

Formulation In ethanol.
Description 97% by TLC
Target Proteins PLA2
CAS 141022-99-3
Molecular Formula C17H31F3O
Formula Weight 308.4g/mol
Solubility DMSO or EtOH
Form Liquid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.