accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 141091-37-4

IUPAC Name: 2-(cyclohex-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(cyclohex-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 208.11
Molecular Formula: C12H21BO2
InChi Key: QNZFUMVTUFOLRT-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CCCCC1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Quantity 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)