Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Cyclohexenylboronic acid pinacol ester, 97%

Catalog No. AC430315000
Change view
Click to view available options
Quantity:
500 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC430315000 500 mg
AC430310010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC430315000 Supplier Thermo Scientific Chemicals Supplier No. 430315000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 141091-37-4 | C12H21BO2 | 208.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141091-37-4
Molecular Formula C12H21BO2
Molecular Weight (g/mol) 208.11
MDL Number MFCD05663845
InChI Key QNZFUMVTUFOLRT-UHFFFAOYSA-N
Synonym 2-cyclohex-1-en-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-cyclohexen-yl-boronic acid pinacol ester, cyclohexene-1-boronic acid pinacol ester, 1-cyclohexen-1-yl-boronic acid pinacol ester, 2-cyclohexenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-cyclohexeneboronic acid, 2-1-cyclohexenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, cyclohexen-1-ylboronic acid, pinacol ester, 2-1-cyclohexen-1-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-cyclohexeneboronic acid pinacol ester
PubChem CID 10932675
IUPAC Name 2-(cyclohexen-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CCCCC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Tan
Boiling Point 125.0°C (0.2 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 500 mg
Formula Weight 208.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Cyclohexenylboronic acid pinacol ester
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.