accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOClNHSHCl

CAS RN 141109-19-5

IUPAC Name: methyl 2-(2-chlorophenyl)-2-{[2-(thiophen-2-yl)ethyl]amino}acetate hydrochloride
Synonyms: methyl 2-(2-chlorophenyl)-2-{[2-(thiophen-2-yl)ethyl]amino}acetate hydrochloride
Molecular Weight (g/mol): 346.27
Molecular Formula: C15H17Cl2NO2S
InChi Key: ZXANKCFSGFEBQW-UHFFFAOYNA-N
SMILES: Cl.COC(=O)C(NCCC1=CC=CS1)C1=CC=CC=C1Cl
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)