Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-N-[2-(2-thienyl)ethyl]-D-phenylglycine methyl ester hydrochloride, 98%

Catalog No. AAH61936MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH61936MD 250 mg
AAH6193603 1 g
AAH6193606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH61936MD Supplier Thermo Scientific Chemicals Supplier No. H61936MD

CAS: 141109-19-5 | C15H17Cl2NO2S | 346.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141109-19-5
Molecular Formula C15H17Cl2NO2S
Molecular Weight (g/mol) 346.27
MDL Number MFCD08458449,MFCD08460122
InChI Key ZXANKCFSGFEBQW-UHFFFAOYNA-N
Synonym methyl 2r-2-2-chlorophenyl-2-2-thiophen-2-yl ethyl amino acetate hydrochloride
PubChem CID 24938549
SMILES Cl.COC(=O)C(NCCC1=CC=CS1)C1=CC=CC=C1Cl

Specifications

IUPAC Name methyl 2-(2-chlorophenyl)-2-{[2-(thiophen-2-yl)ethyl]amino}acetate hydrochloride
Quantity 250 mg
Formula Weight 346.27
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Chloro-N-[2-(2-thienyl)ethyl]-D-phenylglycine methyl ester hydrochloride
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.