accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOONHSFOONH2F

CAS RN 141286-78-4

IUPAC Name: 2-{4-[(2-benzenesulfonamidoethyl)sulfanyl]-2,6-difluorophenoxy}acetamide
Synonyms: pepa (drug)
Molecular Weight (g/mol): 402.43
Molecular Formula: C16H16F2N2O4S2
InChi Key: GTACSIONMHMRPD-UHFFFAOYSA-N
SMILES: NC(=O)COC1=C(F)C=C(SCCNS(=O)(=O)C2=CC=CC=C2)C=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results