accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3OOOHOOCH3CH3O

CAS RN 14131-84-1

IUPAC Name: 6-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-tetrahydro-2H-furo[3,4-d][1,3]dioxol-4-ol
Synonyms: 6-(2,2-dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyl-tetrahydrofuro[3,4-d][1,3]dioxol-4-ol
Molecular Weight (g/mol): 260.29
Molecular Formula: C12H20O6
InChi Key: JWWCLCNPTZHVLF-UHFFFAOYNA-N
SMILES: CC1(C)OCC(O1)C1OC(O)C2OC(C)(C)OC12
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (3)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)