accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2OH3C(R)N(R)OHOONNH2H2NNHSOOCH3NH

CAS RN 141396-28-3

IUPAC Name: (2R,4R)-1-{5-[(diaminomethylidene)amino]-2-(3-methyl-1,2,3,4-tetrahydroquinoline-8-sulfonamido)pentanoyl}-4-methylpiperidine-2-carboxylic acid hydrate
Synonyms: argatroban hydrate
Molecular Weight (g/mol): 526.65
Molecular Formula: C23H38N6O6S
InChi Key: AIEZTKLTLCMZIA-LINCPPCXSA-N
SMILES: O.C[C@@H]1CCN([C@H](C1)C(O)=O)C(=O)C(CCCN=C(N)N)NS(=O)(=O)C1=CC=CC2=C1NCC(C)C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results