accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14152-97-7

H3COOONCSOCH3OOCH3OOCH3O

CAS RN 14152-97-7

IUPAC Name: [3,4,5-tris(acetyloxy)-6-isothiocyanatooxan-2-yl]methyl acetate
Synonyms: [3,4,5-tris(acetyloxy)-6-isothiocyanatooxan-2-yl]methyl acetate
Molecular Weight (g/mol): 389.38
Molecular Formula: C15H19NO9S
InChi Key: WOWNQYXIQWQJRJ-UHFFFAOYNA-N
SMILES: CC(=O)OCC1OC(N=C=S)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results