Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,6-Tetra-O-acetyl-beta-D-glucopyranosyl Isothiocyanate 98.0+%, TCI America™
SDP

Catalog No. A55141G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A55141G 1 g
A5514100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A55141G Supplier TCI America Supplier No. A55141G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 14152-97-7 | C15H19NO9S | 389.38 g/mol

Chemical Identifiers

CAS 14152-97-7
Molecular Formula C15H19NO9S
Molecular Weight (g/mol) 389.38
MDL Number MFCD00043085
InChI Key WOWNQYXIQWQJRJ-UHFFFAOYNA-N
Synonym GITC
PubChem CID 99462
IUPAC Name [3,4,5-tris(acetyloxy)-6-isothiocyanatooxan-2-yl]methyl acetate
SMILES CC(=O)OCC1OC(N=C=S)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O

Specifications

Melting Point 116°C
Color White
Quantity 1 g
Formula Weight 389.38
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,4,6-Tetra-O-acetyl-beta-D-glucopyranosyl Isothiocyanate [for HPLC Labeling]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.