accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3FFNCH3H3C

CAS RN 141546-96-5

IUPAC Name: ethyl 3-(dimethylamino)-3-ethoxy-2,2-difluoropropanoate
Synonyms: ethyl 3-(dimethylamino)-3-ethoxy-2,2-difluoropropanoate
Molecular Weight (g/mol): 225.24
Molecular Formula: C9H17F2NO3
InChi Key: WQKRWROIDBGNCS-UHFFFAOYNA-N
SMILES: CCOC(N(C)C)C(F)(F)C(=O)OCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results