Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl 3-(Dimethylamino)-3-ethoxy-2,2-difluoropropionate 95.0+%, TCI America™
SDP

Catalog No. D27825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27825G 5 g
D27821G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D27825G Supplier TCI America Supplier No. D27825G

Chemical Identifiers

CAS 141546-96-5
Molecular Formula C9H17F2NO3
Molecular Weight (g/mol) 225.236
MDL Number MFCD00722253
InChI Key WQKRWROIDBGNCS-UHFFFAOYSA-N
Synonym 3-(Dimethylamino)-3-ethoxy-2,2-difluoropropionic Acid Ethyl Ester
PubChem CID 2758500
IUPAC Name ethyl 3-(dimethylamino)-3-ethoxy-2,2-difluoropropanoate
SMILES CCOC(C(C(=O)OCC)(F)F)N(C)C

Specifications

Color Yellow
Boiling Point 62°C
Quantity 5 g
Formula Weight 225.24
Percent Purity ≥95.0% (N)
Physical Form Liquid
Chemical Name or Material Ethyl 3-(Dimethylamino)-3-ethoxy-2,2-difluoropropionate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.