accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNNCH3CH3CCH3CH3

CAS RN 141556-42-5

IUPAC Name: 1,3-bis(2,4,6-trimethylphenyl)-3H-1λ⁵-imidazol-1-ylium-2-ide
Synonyms: IMes
Molecular Weight (g/mol): 304.44
Molecular Formula: C21H24N2
InChi Key: JCYWCSGERIELPG-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(N2C=C[N+](=[C-]2)C2=C(C)C=C(C)C=C2C)C(C)=C1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results