accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NNClO

CAS RN 14160-93-1

IUPAC Name: 4-amino-6-chloropyrimidine-5-carbaldehyde
Synonyms: 4-amino-6-chloropyrimidine-5-carbaldehyde
Molecular Weight (g/mol): 157.56
Molecular Formula: C5H4ClN3O
InChi Key: GOJNFUXEBVBARW-UHFFFAOYSA-N
SMILES: NC1=NC=NC(Cl)=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results