accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14167-18-1

IUPAC Name: λ²-cobalt(2+) 2-{[(2-{[(2-oxidophenyl)methylidene]amino}ethyl)imino]methyl}benzen-1-olate
Synonyms: salcomine
Molecular Weight (g/mol): 325.23
Molecular Formula: C16H14CoN2O2
InChi Key: NPAQSKHBTMUERN-UHFFFAOYSA-L
SMILES: [Co++].[O-]C1=CC=CC=C1C=NCCN=CC1=CC=CC=C1[O-]
Molecular Weight (g/mol) 
  • (7)
  • (1)
Quantity 
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (4)
  • (7)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
  • (4)