Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Mono-Bromoborane Methyl Sulfide Complex Solution

Catalog No. 262064100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-206-4100ML 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 26-206-4100ML Supplier Sigma Aldrich Supplier No. 262064100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactant for:Substitution reactions for synthesis of N-heterocyclic carbene boranes with boron-heteroatom bondsHydroboration reactionsRegio- and chemoselective brominative cleavage of terminal epoxides

Specifications

CAS 14167-18-1
Molecular Formula C16H14CoN2O2
Linear Formula [-CH2N=CHC6H4-2-(O-)]2Co
MDL Number MFCD00000009
Quantity 1 g
Synonym Bromo(dimethyl sulfide)borane; Bromo(dimethyl sulfide)dihydroboron
Molecular Weight (g/mol) 325.23 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.