accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3

CAS RN 14167-59-0

IUPAC Name: tetratriacontane
Synonyms: N-tetratriacontane
Molecular Weight (g/mol): 478.93
Molecular Formula: C34H70
InChi Key: GWVDBZWVFGFBCN-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
Percent Purity 
  • (3)
  • (2)
  • (1)
Physical Form 
  • (5)
  • (1)
Quantity 
  • (3)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (5)
  • (1)
  • (3)
  • (2)
  • (1)
  • (5)
  • (1)
  • (3)
  • (1)
  • (2)
  • (5)