accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14174-08-4

IUPAC Name: 2,3,5,6,8,9-hexahydro-1,4,7,10-benzotetraoxacyclododecine
Synonyms: 2,3,5,6,8,9-hexahydro-1,4,7,10-benzotetraoxacyclododecine
Molecular Weight (g/mol): 224.26
Molecular Formula: C12H16O4
InChi Key: OAJNZFCPJVBYHB-UHFFFAOYSA-N
SMILES: C1COCCOC2=CC=CC=C2OCCO1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results