accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14187-32-7

IUPAC Name: 2,5,8,15,18,21-hexaoxatricyclo[20.4.0.0⁹,¹⁴]hexacosa-1(26),9,11,13,22,24-hexaene
Synonyms: dibenzo-18-crown-6 polyether XXVIII crown 18 dibenzo(b,k)(1,4,7,10,13,16)hexaoxacyclooctadecane dicyclohexano-18-crown-6 dibenzo-18-crown-6-ether
Molecular Weight (g/mol): 360.41
Molecular Formula: C20H24O6
InChi Key: YSSSPARMOAYJTE-UHFFFAOYSA-N
SMILES: C1COC2=CC=CC=C2OCCOCCOC2=CC=CC=C2OCCO1
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (2)
  • (4)
  • (2)
  • (3)
  • (4)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)