accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COO(R)OHN

CAS RN 141942-85-0

IUPAC Name: ethyl (3R)-4-cyano-3-hydroxybutanoate
Synonyms: ethyl (3R)-4-cyano-3-hydroxybutanoate
Molecular Weight (g/mol): 157.17
Molecular Formula: C7H11NO3
InChi Key: LOQFROBMBSKWQY-ZCFIWIBFSA-N
SMILES: CCOC(=O)C[C@H](O)CC#N
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (5)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)