accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3CH2

CAS RN 142-09-6

IUPAC Name: hexyl 2-methylprop-2-enoate
Synonyms: hexyl methacrylate
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: LNCPIMCVTKXXOY-UHFFFAOYSA-N
SMILES: CCCCCCOC(=O)C(C)=C
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)