Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Hexyl methacrylate, 97%, stab. with 100ppm 4-methoxyphenol

Catalog No. AAL1156922
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1156922 100 g
AAL1156914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1156922 Supplier Thermo Scientific Chemicals Supplier No. L1156922
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 142-09-6 | C10H18O2 | 170.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-09-6
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD00015283
InChI Key LNCPIMCVTKXXOY-UHFFFAOYSA-N
Synonym hexyl methacrylate, n-hexyl methacrylate, hexyl 2-methyl-2-propenoate, methacrylic acid, hexyl ester, 2-propenoic acid, 2-methyl-, hexyl ester, unii-538b2q3hsv, ccris 4824, hexyl 2-methylacrylate, methacrylic acid hexyl ester, dsstox_cid_5406
PubChem CID 8872
IUPAC Name hexyl 2-methylprop-2-enoate
SMILES CCCCCCOC(=O)C(C)=C

Specifications

Melting Point -57°C
Density 0.885
Boiling Point 204°C
Flash Point 80°C (176°F)
Odor Characteristic
Refractive Index 1.431
Quantity 100 g
Beilstein 1754703
Formula Weight 170.25
Percent Purity 97%
Chemical Name or Material n-Hexyl methacrylate, stabilized with 100ppm 4-methoxyphenol
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 205-521-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.