CAS RN 142-19-8
IUPAC Name:
prop-2-en-1-yl heptanoate
Synonyms:
allyl heptanoate
Molecular Weight (g/mol):
170.25
Molecular Formula:
C10H18O2
InChi Key:
SJWKGDGUQTWDRV-UHFFFAOYSA-N
SMILES:
CCCCCCC(=O)OCC=C