Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl Heptanoate 98.0+%, TCI America™
SDP

Catalog No. A211925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A211925G 25 g
A2119250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A211925G Supplier TCI America Supplier No. A211925G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 142-19-8
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD00038341
InChI Key SJWKGDGUQTWDRV-UHFFFAOYSA-N
Synonym allyl heptanoate, allyl heptylate, 2-propenyl heptanoate, allyl enanthate, allyl heptoate, heptanoic acid, 2-propenyl ester, prop-2-en-1-yl heptanoate, heptanoic acid, allyl ester, allylester kyseliny enanthove, heptanoic acid, 2-propen-1-yl ester
PubChem CID 8878
IUPAC Name prop-2-enyl heptanoate
SMILES CCCCCCC(=O)OCC=C

Specifications

Melting Point -66°C
Color Yellow
Boiling Point 89°C
Quantity 25 g
UN Number 2810
Formula Weight 170.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Heptanoate
EINECSNumber (2)-0759
RTECSNumber MJ1750000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.