accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCl

CAS RN 142-28-9

IUPAC Name: 1,3-dichloropropane
Synonyms: dichloropropane
Molecular Weight (g/mol): 112.98
Molecular Formula: C3H6Cl2
InChi Key: YHRUOJUYPBUZOS-UHFFFAOYSA-N
SMILES: ClCCCCl
Molecular Weight (g/mol) 
  • (4)
  • (4)
Boiling Point 
  • (2)
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (4)
  • (1)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (5)
  • (2)
  • (1)