Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dichloropropane, 99%

Catalog No. AC113682500
Change view
Click to view available options
Quantity:
50 mL
250 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC113682500 250 mL Glass bottle
AC113680500 50 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC113682500 Supplier Thermo Scientific Chemicals Supplier No. 113682500
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 142-28-9 | C3H6Cl2 | 112.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-28-9
Molecular Formula C3H6Cl2
Molecular Weight (g/mol) 112.99
MDL Number MFCD00000999
InChI Key YHRUOJUYPBUZOS-UHFFFAOYSA-N
Synonym trimethylene dichloride, propane, 1,3-dichloro, trimethylene chloride, 1,3-dichloro-propane, unii-aj1hq2gucp, ccris 9220, ch2clch2ch2cl, aj1hq2gucp, cl ch2 3cl, 1,3-dichlorpropan
PubChem CID 8881
IUPAC Name 1,3-dichloropropane
SMILES C(CCl)CCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -99.0°C
Color Colorless
Density 1.1900g/mL
Boiling Point 120.0°C to 122.0°C
Flash Point 20°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4471 to 1.4491
Linear Formula Cl(CH2)3Cl
Quantity 250 mL
Beilstein 01, 105
Specific Gravity 1.19
Solubility Information Solubility in water: 0.8g/L (20°C). Other solubilities: freely soluble in ethanol and ether
Viscosity 1.1 mPa.s (20°C)
Formula Weight 112.99
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material 1, 3-Dichloropropane, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-531-3
RTECSNumber TX9660000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.