accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOHOO

CAS RN 142-45-0

IUPAC Name: but-2-ynedioic acid
Synonyms: acetylenedicarboxylic acid
Molecular Weight (g/mol): 114.06
Molecular Formula: C4H2O4
InChi Key: YTIVTFGABIZHHX-UHFFFAOYSA-N
SMILES: OC(=O)C#CC(O)=O
Percent Purity 
  • (2)
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (4)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (4)
  • (3)
  • (2)
  • (2)