Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acetylenedicarboxylic acid, 98%

Catalog No. AC102521000
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC102521000 100 g Glass bottle
AC102520250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC102521000 Supplier Thermo Scientific Chemicals Supplier No. 102521000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 142-45-0 | C4H2O4 | 114.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-45-0
Molecular Formula C4H2O4
Molecular Weight (g/mol) 114.06
MDL Number MFCD00004362
InChI Key YTIVTFGABIZHHX-UHFFFAOYSA-N
Synonym acetylenedicarboxylic acid, 2-butynedioic acid, butynedioic acid, acetylenedicarboxylate, unii-2d2oj4ko44, acetylenedicarboxylic acid 8ci, acetylendicarboxylate, hoocc=ccooh, acetylenedicarboxylicacid, pubchem13735
PubChem CID 371
ChEBI CHEBI:30781
IUPAC Name but-2-ynedioic acid
SMILES OC(=O)C#CC(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 180°C to 187°C
Color Beige to Cream or White
Flash Point 131°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HO2CC∽CCO2H
Quantity 100 g
Beilstein 02, 801
Fieser 01,12
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and ether
Formula Weight 114.06
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Acetylenedicarboxylic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Toxic if swallowed.
Toxic in contact with skin.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 205-536-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.