accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 142-68-7

IUPAC Name: oxane
Synonyms: tetrahydropyran THP
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: DHXVGJBLRPWPCS-UHFFFAOYSA-N
SMILES: C1CCOCC1
Molecular Weight (g/mol) 
  • (1)
  • (8)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (3)
  • (5)
  • (1)
  • (8)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (1)