Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydropyran, 98+%

Catalog No. AAA13392AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA13392AP 500 mL
AAA13392AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA13392AP Supplier Thermo Scientific Chemicals Supplier No. A13392AP
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 142-68-7 | C5H10O | 86.134 g/mol

It is is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuff. They are also used as important solvents, as chemical intermediate and as monomer for ring-opening polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuff. They are also used as important solvents, as chemical intermediate and as monomer for ring-opening polymerization.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Oxidizing agents, Strong acids. May form peroxides in the absence of inhibitors. Light sensitive
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-68-7
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.134
MDL Number MFCD00006585
InChI Key DHXVGJBLRPWPCS-UHFFFAOYSA-N
Synonym tetrahydropyran, tetrahydro-2h-pyran, oxacyclohexane, pentamethylene oxide, 2h-pyran, tetrahydro, tetrahydropyrane, unii-v06i3ilg6b, 2h-tetrahydropyran, v06i3ilg6b, pye
PubChem CID 8894
ChEBI CHEBI:46941
IUPAC Name oxane
SMILES C1CCOCC1

Specifications

Melting Point -49°C
Density 0.882
Boiling Point 87°C to 89°C
Flash Point −15°C (5°F)
Refractive Index 1.421
Quantity 500 mL
UN Number UN3271
Beilstein 102436
Merck Index 14,9217
Solubility Information It is soluble in water.
Formula Weight 86.14
Percent Purity ≥98%
Chemical Name or Material Tetrahydropyran
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
EINECSNumber 205-552-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.