accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 142-75-6

IUPAC Name: 2-hydroxypropyl octadecanoate
Synonyms: propylene glycol monostearate
Molecular Weight (g/mol): 342.56
Molecular Formula: C21H42O3
InChi Key: FKOKUHFZNIUSLW-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OCC(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results