Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Propylene Glycol Monostearate, MP Biomedicals

Catalog No. ICN218521 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN218521 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN218521 Supplier MP Biomedicals, Inc Supplier No. 0521852180

Chemical Identifiers

CAS 142-75-6
Molecular Formula C21H42O3
Molecular Weight (g/mol) 342.564
InChI Key FKOKUHFZNIUSLW-UHFFFAOYSA-N
Synonym 2-hydroxypropyl stearate, propylene glycol monostearate, propylene glycol stearate, monosteol, prostearin, pegosperse ps, monosteol tg, cerasynt pa, cerasynt pn, dragil-p
PubChem CID 14878
IUPAC Name 2-hydroxypropyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(C)O

Specifications

Quantity 100 g
Formula Weight 342.63
Chemical Name or Material Propylene Glycol Monostearate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.