accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 142-81-4

IUPAC Name: hydrogen hexan-1-amine chloride
Synonyms: hydrogen hexylamine chloride
Molecular Weight (g/mol): 137.65
Molecular Formula: C6H16ClN
InChi Key: XKDUZXVNQOZCFC-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCCCCCN
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results