Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hexylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. H013525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H013525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H013525G Supplier TCI America Supplier No. H013525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 142-81-4
Molecular Formula C6H16ClN
Molecular Weight (g/mol) 137.65
MDL Number MFCD00043792
InChI Key XKDUZXVNQOZCFC-UHFFFAOYSA-N
Synonym 1-Aminohexane Hydrochloride
PubChem CID 67342
IUPAC Name hydrogen hexan-1-amine chloride
SMILES [H+].[Cl-].CCCCCCN

Specifications

Melting Point 223°C
Color White
Quantity 25 g
Formula Weight 137.65
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material Hexylamine Hydrochloride
EINECSNumber (2)-0133
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.