CAS RN 142036-63-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ PPACK, Dihydrochloride
PPACK, Dihydrochloride, CAS 142036-63-3, is an extremely potent and selective irreversible inhibitor of thrombin (Kobs/[I] = 107M-1S-1). Reacts with thrombin in a 1:1 stoichiometry.
2
MilliporeSigma™ Calbiochem™ InSolution™ PPACK, 2HCl
A 25 mM (5 mg/357 μl) sterile-filtered solution of PPACK, 2HCl in H2O.