accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 14212-87-4

IUPAC Name: 4-[1-(morpholin-4-yl)ethenyl]morpholine
Synonyms: 4-[1-(morpholin-4-yl)ethenyl]morpholine
Molecular Weight (g/mol): 198.27
Molecular Formula: C10H18N2O2
InChi Key: MNEZCVVWTRIDOE-UHFFFAOYSA-N
SMILES: C=C(N1CCOCC1)N1CCOCC1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results