accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CNCH3CH3SOOBr

CAS RN 1421261-62-2

IUPAC Name: 2-bromo-N,N-bis(propan-2-yl)benzene-1-sulfonamide
Synonyms: 2-bromo-N,N-diisopropylbenzenesulfonamide
Molecular Weight (g/mol): 320.25
Molecular Formula: C12H18BrNO2S
InChi Key: OCXVEJYCGJCHCW-UHFFFAOYSA-N
SMILES: CC(C)N(C(C)C)S(=O)(=O)C1=CC=CC=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results