accessibility menu, dialog, popup

UserName

Viewing profile as user:
SOONHNOOBr

CAS RN 1421261-66-6

IUPAC Name: N-(2-bromo-5-nitrophenyl)benzenesulfonamide
Synonyms: N-(2-bromo-5-nitrophenyl)benzenesulfonamide
Molecular Weight (g/mol): 357.18
Molecular Formula: C12H9BrN2O4S
InChi Key: KTKONBLIZGUGBA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(NS(=O)(=O)C2=CC=CC=C2)=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results